Hanging Cell Models
Product 470135-946 is similar to 470000-002 but manufacturer and specifications may vary.
Product Details & Documents
For Demonstration And Display
These high visibility large models are vacuum formed sheets of heavy plastic, molded to show in relief all the known organelles and inclusions of these two important cells. Twenty structures are included in the animal cell; twenty-three in the plant cell. All are identified by the accompanying key cards. Hand painted in brilliant color and are fitted with an easel back which permits the model to sit upright on a surface of hang on a wall with equal ease.
Specifications
- Description:Animal Cell
- Length:46 cm
- No. of Keyed Items:20
- Width:61 cm
- No. of Parts:1
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



