VWR® Basic SPP Lab Coats
Product Details & Documents
Disposable lab coats made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Designed for comfort and contamination control, these lab coats are ideal for clean areas, food processing, and general industrial use.
Spunbond Polypropylene (SPP) material is lightweight and breathable
- Breathable fabric reduces heat buildup during extended use
- Snap front closure for easy donning and removal
- Available with elastic or knit cuff styles
- Non-linting fabric helps maintain cleanliness in controlled environments
- Single-use, disposable design for contamination control
- Economical solution for basic protection needs
- Disposable design simplifies protocols and reduces laundering costsSpunbond Polypropylene (SPP) material is lightweight and breathable
- Breathable fabric reduces heat buildup during extended use
- Snap front closure for easy donning and removal
- Available with elastic or knit cuff styles
- Non-linting fabric helps maintain cleanliness in controlled environments
- Single-use, disposable design for contamination control
- Economical solution for basic protection needs
- Disposable design simplifies protocols and reduces laundering costs
The VWR® Basic Protection Lab Coat is designed for environments where contamination control and user comfort are essential, but exposure risks remain minimal. Constructed from spunbond polypropylene (SPP), this nonwoven fabric offers a lightweight, breathable solution that promotes airflow while maintaining cleanliness. The non-linting properties help reduce particle contamination, making it suitable for clean areas, food processing, and general industrial applications.
The lab coat features a snap-front closure for quick and secure fastening, along with elastic or knit cuffs to accommodate different glove types and ensure a comfortable fit. The disposable design supports hygiene compliance by reducing cross-contamination risks and simplifying inventory management. While not intended for hazardous chemical or liquid protection, this garment provides an effective barrier against dust, dirt, and dry particulates, making it ideal for routine tasks in controlled environments.
By combining economical single-use convenience, breathability, and basic contamination control, VWR® Basic Protection SPP Lab Coats deliver a practical solution for facilities seeking dependable protective apparel for everyday operations.
Specifications
- Clothing Size:2XL
- Color:White
- Length:43½"
- Chest Size:29½"
- Sleeve:37½"
- Cat. No.:89136-824
- Cleanroom class:10000/ISO 7
- Sleeve or leg length:Long
- Garment application:Cleanroom|Industrial|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Fabric:Spunbond polypropylene
- Particle shedding (Helmke Drum):Level II
- Water resistance:First line barrier
- Weight:40 g/m²
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



