Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Registrarse
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COVER GLASS OCTAG 20X26MM PK12
Catalog # 15170-296
Proveedor:
Erie Scientific
Expand image
COVER GLASS OCTAG 20X26MM PK12
Catalog # 15170-296
Proveedor:
Erie Scientific
Supplier Number:
2026O
Frequently Bought Together
VWR® Disposable Serological Pipettes
Proveedor: VWR International
VWR® Disposable Petri Dishes
Proveedor: VWR International
VWR® Nitrile Examination Gloves, Blue
Proveedor: VWR International
VWR® Universal Pipette Tips
Proveedor: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Proveedor: BDH