Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Peptone

VWR® Peptone

Peptone, 1 kg
Catalog # 97063-324
Supplier:   VWR
Supplier #:  J636-1KG
VWR® Peptone
Catalog # 97063-324
Supplier:  VWR
CAS Number:  
VWR® Peptone
VWR® Peptone

Peptone, 1 kg

CAS Number:  
Price
$5,352.06
Log in for availability
Each (1kg) Case de 4 (1kg)
Product Details & Documents

High purity protein digests used for media supplements.

  • High purity
  • Used for media supplements

VWR® Life Science VWR® Peptone is a high purity protein used for media supplements.

Documents

Specifications

Specifications

  • Description:
    Peptone
  • Size:
    1 kg
  • Application:
    An enzymatic digestion of protein

Specifications