Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® TRIS Buffer, 1 M Solution, pH 8.0

VWR® TRIS Buffer, 1 M Solution, pH 8.0

500 ml
Catalog # 97062-674
Supplier:   VWR
Número de proveedor #:  E199-500ML
VWR® TRIS Buffer, 1 M Solution, pH 8.0
Catalog # 97062-674
Supplier:  VWR
CAS Number:  
VWR® TRIS Buffer, 1 M Solution, pH 8.0
VWR® TRIS Buffer, 1 M Solution, pH 8.0

500 ml

CAS Number:  
Price
$1,477.98
Log in Para disponibilidad
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tris (hydroxymethyl) aminomethane

  • Biotechnology grade
  • pH @25C 7.9 - 8.1

Storage conditions: Room temperature

Documents

Especificaciones

Especificaciones

  • Size:
    500 ml

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.