D-(-)-Fructose, powder, high purity
Bottle, 1 kg
Supplier: VWR
: 0226-1KG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Fine white powder.
Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
- CAS Number:57-48-7
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00148910
- Molecular Weight:180.16 g/mol
- Merck Index:13,04295
- Boiling Pt:552 °C (1013 hPa)
- Melting Pt:100 - 110 °C
- Melting Pt:100…110 °C
- Density:1.59 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-333-3
Specifications
Specification Test Results
- Arsenic:= 0.0001%
- Calcium & Magnesium:= 0.005%
- Chloride:= 0.018%
- Heavy Metals (as Pb):= 0.0005%
- Loss on Drying:= 0.5%
- Purity:= 99%
- Residue on Ignition:= 0.5%
- Sulfate:= 0.025%
Specifications
- Pack type:Bottle
- Size:1 kg



