Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Purple
Catalog # 24270-118
Supplier #:  375922
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # 24270-118
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Purple
Price
$2,155.19
Log in for availability
Paquete de 500 Case de 4 (500UN)
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
  • Made of polystyrene
  • Available in 10 different colors
Ordering information: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Specifications
  • Color:
    Purple