Eppendorf® Safe-Lock Tubes, 2.0 ml - Microtubes
Product Details & Documents
Eppendorf® Safe-Lock 2.0 ml microtube ensure secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2.0 ml capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.
- Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage
- g-Safe® technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
- g-Safe® technology prevents sample loss from tube breakage, and enhances safety when handling hazardous samples
- Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures
- Precise lid sealing minimizes evaporation rates during prolonged storage
- Large frosted lid and side surface for easy labeling
- Functionality is assured within a temperature range from −86 to +100 °C
- Autoclavable when open (121 °C, 20 min.) for effective sterilization
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity
Eppendorf® Safe-Lock 2.0 ml microtubes redefine secure sample storage. The patented Safe-Lock mechanism ensures protection against accidental openings, preserving sample integrity meticulously. Versatile across laboratory applications, the 2.0 ml capacity adapts to diverse sample volumes. Constructed for durability, these microtubes withstand rigorous daily use. The user-friendly design, featuring clear graduations and a smooth surface, enhances visibility and handling ease. Elevate laboratory efficiency with Eppendorf® Safe-Lock tubes - trusted for reliability, security, and performance in sample management.
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Safe-Lock
Material: PP
Volume: 2.0 ml
Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.
Specifications
- Color:Colorless
- Packaging:500 tubes (1 bag×500 tubes)
- Maximum RCF:25000×g
- Cat. No.:20901-540
- Temperature range:−86…+100 °C
- Autoclavable:When open 121 °C, 20 mins
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



