Sodium hydroxide, pellets, Proteomics Grade
HDPE bottle, 500 g
Supplier: VWR
Supplier #: M137-500G
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390°C
- Boiling Pt:1390 °C
- Melting Pt:318°C
- Melting Pt:318 °C
- Density:2.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
Specifications
Specification Test Results
- Chloride:0.005%
- DNase:none detected
- Heavy Metals:0.002 %
- Iron:= 0.001 %
- Mercury:= 0.1 ppm
- Nickel:= 0.001 %
- Nitrogen Compounds:= 0.001 %
- Phosphate:= 0.001 %
- Potassium:= 0.02 %
- Protease:none
- Purity:= 97 %
- RNase:none detected
- Sodium Carbonate:= 1 %
- Sulfate:= 0.003%
Specifications
- Pack type:HDPE bottle
- Size:500 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



