Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Peptone

VWR® Peptone

Peptone 140, 1 kg
Catalog # 97064-364
VWR® Peptone
Catalog # 97064-364
Supplier:  
CAS Number:  
VWR® Peptone
VWR® Peptone

Peptone 140, 1 kg

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MEDIUM PEPTONE BACT CNTS HI TRP 500G Catalog #61001-506

Product 61001-506 is a replacement for 97064-364 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

High purity protein digests used for media supplements.

  • High purity
  • Used for media supplements

VWR® Life Science VWR® Peptone is a high purity protein used for media supplements.

Documents

Specifications

Specifications

  • Description:
    Peptone 140
  • Size:
    1 kg
  • Application:
    Hydrolysate digest of soybean

Specifications