Order Entry
Mexico
Orders LinkContactUsLinkComponent
Agar for bacteriology

Agar for bacteriology

1 kg
Catalog # 97064-332
Supplier:   VWR
Supplier #:  J637-1KG
Agar for bacteriology
Catalog # 97064-332
Supplier:  VWR
CAS Number:  9002-18-0
Agar for bacteriology
Agar for bacteriology

1 kg

CAS Number:  9002-18-0
Price
$4,686.14
Log in for availability
Each (1UN) Case de 4 (1UN)
Product Details & Documents

A solidifying agent used in bacteriology. Agar consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.

  • Agar for the growth of bacteria and fungi
  • Typically used at concentrations of 1-2% in solid media preparation
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Specifications

Specifications

  • Size:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.