Urea, high purity
Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions. Effectively denatures RNA. For use in nucleic electrophoresis applications.
- :57-13-6
- :NH₂CONH₂
- :MFCD00008022
- :60.06 g/mol
- :13,09935
- :197 °C (1013 hPa)
- :132.7 °C
- :1.34 g/cm³ (20 °C)
- :Ambient
- :200-315-5
- :01-2119463277-33
- Abs.@280nm (5M, Water):0.05
- Biuret Test:Pass
- Chloride:= 0.0005%
- Cyanate:= 0.002%
- DNase:none detected
- Heavy Metals (as Pb):= 0.0005%
- Insolubles:= 0.005%
- Iron:= 0.0001%
- Melting Point:132°C - 135°C
- Protease:none detected
- Purity:= 99.5%
- RNase:none detected
- Solubility (5M, Water):Pass
- Size:5 kg
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



