Order Entry
Mexico
Orders LinkContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table
Bottle, 500 ml, 7.00
  97007-786
Supplier #:  63056125
NIST Traceable pH Buffers with Temperature Table
  97007-786
 :  
 :  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table
Bottle, 500 ml, 7.00
 :  
NIST-traceable buffer for pH electrode calibration and verification.
  • Temperature table on bottle
Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.
Documents
  • Pack type:
    Bottle
  • Size:
    500 ml
  • pH:
    7.00