Order Entry
Mexico
Orders LinkContactUsLinkComponent
Prefilter for Masks, 3M™
Prefilter for Masks, 3M™
P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators., Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
Catalog # 56222-931
Marketsource
Supplier #:  2091
Prefilter for Masks, 3M™
Catalog # 56222-931
Marketsource
Prefilter for Masks, 3M™
P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators., Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
Price
$15,360.00
Log in for availability
Paquete de 2 Case de 50 (2UN)
Product Details & Documents
For use with 5000-series respirators.
  • NIOSH 42 CFR 84 approved
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Specifications
  • Description:
    P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators.
  • Approved For Use Against:
    Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators