Order Entry
Mexico
Orders LinkContactUsLinkComponent
epT.I.P.S.® Bulk - Pipette Tips

epT.I.P.S.® Bulk - Pipette Tips

500 - 10000 µl, 243 mm, Turquoise, 200 Tips (2 bags×100 Tips)
Catalog # 47747-992
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   EPPENDORF BIO TOOLS
Supplier #:  022492101
epT.I.P.S.® Bulk - Pipette Tips
Catalog # 47747-992
Product available on GSA Advantage®
   Low Manufacturing Impact
epT.I.P.S.® Bulk - Pipette Tips
epT.I.P.S.® Bulk - Pipette Tips

500 - 10000 µl, 243 mm, Turquoise, 200 Tips (2 bags×100 Tips)

Price
$2,810.00
Log in for availability
Case (200UN)
Product Details & Documents

Eppendorf epT.I.P.S.® Bulk packs offer a practical, high-volume solution with 'Eppendorf Quality' pipette tips, ensuring precise and reliable measurements for various lab applications, packaged economically for high-throughput settings.

  • Original, high-quality Eppendorf pipette tips packaged in resealable bags
  • Available in sizes from 10 µl to 10 ml in Eppendorf Quality purity grade
  • epT.I.P.S.® Standard 200, 300 and 1000 µl tips are also available color-coded yellow and blue
  • epT.I.P.S.® Standard pipette tips can be autoclaved at 121 °C in 20 min
  • Fine retracted or extended tips simplify work with small volumes
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally integrated pipetting system') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

Certifications: General quality and ISO certificates available.

Documents

Specifications
  • Volume:
    500 - 10000 µl
  • Length:
    243 mm
  • Packed:
    200 Tips (2 bags×100 Tips)
  • Color Code:
    Turquoise
  • Cat. No.:
    47747-992
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact