Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes
Product Details & Documents
Eppendorf® Safe-Lock 1.5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1.5 ml capacity.
- Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
- g-Safe® technology, depending on the Safe-Lock Tube variant (refer to the manual), ensures outstanding centrifugation stability up to 30000×g
- g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
- Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
- Precision lid sealing minimizes evaporation rates during extended storage periods
- Generously sized frosted lid and side surface facilitate easy labeling
- Assured functionality in a temperature range from −86 to +100 °C
- Autoclavable when open (121 °C, 20 min.) for reliable sterilization
- Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity
Eppendorf® Safe-Lock 1.5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1.5 ml capacity cater to varied sample volumes. Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock tubes.
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Safe-Lock
Material: PP
Volume: 1.5 ml
Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.
Documents
Certificates
Search by Lot NumberSpecifications
- Color:Colorless
- Packaging:500 tubes (1 bag×500 tubes)
- Maximum RCF:30000×g
- Cat. No.:21008-959
- Temperature range:−86…+100 °C
- Autoclavable:When open 121 °C, 20 mins



