Inoculum Broth, BD
B12 Inoculum Broth, 100 g
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Supplier #: 254210
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.
- Used for cultivating lactobacilli
- For preparing the inoculum for microbiological assays
- Peptone provides nitrogen and amino acids
- Beige, homogenous appearance when dehydrated
Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:B12 Inoculum Broth
- Size:100 g