Order Entry
Mexico
Orders LinkContactUsLinkComponent
epT.I.P.S.® Singles, Pipette Tips

epT.I.P.S.® Singles, Pipette Tips

2 - 200 µl, +, 53 mm, Yellow, 1,000 Tips, individually Packed
  47745-218
Supplier #:  022492209
epT.I.P.S.® Singles, Pipette Tips
  47745-218
epT.I.P.S.® Singles, Pipette Tips

2 - 200 µl, +, 53 mm, Yellow, 1,000 Tips, individually Packed

 
$5,450.00
   
Case (1,000UN)
 

Eppendorf epT.I.P.S.® Singles high-precision pipette tips offer unparalleled precision in 'Biopur®' and 'Sterile' grades, featuring individually wrapped tips for maximum sterility. Ideal for sensitive and critical lab applications where contamination control is paramount.

  • Eppendorf pipette tip, individually wrapped
  • Available in Biopur and Sterile purity grades
  • For use in medical, pharmaceutical, food industry, molecular biology, and cell technology
  • Batch number and expiration date on each blister package
  • Each production lot inspected continuously by an independent laboratory
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs.

Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

Certifications: Lot-specific certficates for the purity grades Sterile, Biopur® can be downloaded from the Eppendorf website.

Documents

 
  • Volume:
    2 - 200 µl
  • Length:
    53 mm
  • Packed:
    1,000 Tips, individually Packed
  • Sterile:
    +
  • Color Code:
    Yellow