Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Technical Buffer Solutions, Sachets

±0.02, Sachets, blue, 30×20 ml, 10.01
Catalog # 11238-442
 :  51302079
Technical Buffer Solutions, Sachets
Catalog # 11238-442
CAS Number:  
Technical Buffer Solutions, Sachets
Technical Buffer Solutions, Sachets

±0.02, Sachets, blue, 30×20 ml, 10.01

CAS Number:  
Price
$4,569.09
Log in for availability
Paquete de 30
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

NIST-traceable buffer for pH electrode calibration and verification.

  • NIST-traceable
  • Ensure freshness
  • Improve data

Also available in multiple packaging options.

Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.

Specifications

Specifications

  • Pack type:
    Sachets, blue
  • Size:
    30×20 ml
  • pH:
    10.01
  • pH Accuracy:
    ±0.02

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.