Order Entry
Mexico
Orders LinkContactUsLinkComponent
Urea, high purity

Urea, high purity

2.5 kg
Catalog # 97061-922
Supplier:   VWR
Supplier #:  0568-2.5KG
Urea, high purity
Catalog # 97061-922
Supplier:  VWR
CAS Number:  57-13-6
Urea, high purity
Urea, high purity

2.5 kg

CAS Number:  57-13-6
Price
$3,861.63
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions. Effectively denatures RNA. For use in nucleic electrophoresis applications.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specification Test Results

  • Abs.@280nm (5M, Water):
    0.05
  • Biuret Test:
    Pass
  • Chloride:
    = 0.0005%
  • Cyanate:
    = 0.002%
  • DNase:
    none detected
  • Heavy Metals (as Pb):
    = 0.0005%
  • Insolubles:
    = 0.005%
  • Iron:
    = 0.0001%
  • Melting Point:
    132°C - 135°C
  • Protease:
    none detected
  • Purity:
    = 99.5%
  • RNase:
    none detected
  • Solubility (5M, Water):
    Pass

Specifications

  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.