Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dimidium bromide

Dimidium bromide

Bottle, 250 mg
Catalog # 220065-689
Irritant
Supplier #:  41785-250MG
Dimidium bromide
Catalog # 220065-689
CAS Number:  518-67-2
Dimidium bromide
Dimidium bromide

Bottle, 250 mg

CAS Number:  518-67-2
Price
$6,703.44
Log in for availability
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
  • CAS Number:518-67-2
  • Formula:C₂₀H₁₈BrN₃
  • MDL Number:MFCD00011757
  • Molecular Weight:380.29 g/mol
  • Melting Pt:243 - 248 °C
  • Melting Pt:243…248 °C
  • Storage Temperature:Ambient
  • EINECS:208-256-7
  • UN:2811
  • ADR:6.1,I

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.