Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Burets, Automatic Self-Zeroing, with Stopcock

VWR® Burets, Automatic Self-Zeroing, with Stopcock

50 ml, 775 mm, 0.10 ml
Catalog # 89012-366
Supplier:   VWR International
VWR® Burets, Automatic Self-Zeroing, with Stopcock
Catalog # 89012-366
Supplier:  VWR International
VWR® Burets, Automatic Self-Zeroing, with Stopcock
VWR® Burets, Automatic Self-Zeroing, with Stopcock

50 ml, 775 mm, 0.10 ml

Price
$6,093.05
Log in for availability
Each (1UN)
Product Details & Documents

Borosilicate glass buret for general industrial applications.

  • Borosilicate glass fabrication
  • Provided with fluoropolymer resin stopcocks

Assembly designed to fit a variety of bottle sizes.

Ordering information: Buret supplied with rubber bulb, 2mm stopcock, connecting tube, bottle, air inlet tube, and rubber stopper.

Specifications
  • Graduation Interval:
    0.10 ml
  • Capacity:
    50 ml
  • Length:
    775 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.