Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
473 ml (16 oz.), 8.9 dia.×8.9H cm (3¹/₂×3¹/₂")
Catalog # 89009-666
Supplier:   VWR International
VWR® HDPE Multipurpose Containers
Catalog # 89009-666
Supplier:  VWR International
VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
473 ml (16 oz.), 8.9 dia.×8.9H cm (3¹/₂×3¹/₂")
Price
$1,750.44
Log in for availability
Case (100UN)
Product Details & Documents
Translucent containers for the storage of pathology, industrial, and environmental samples. Containers are made of high quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800mL container is equipped with a handle for portability.
  • HDPE construction
  • Chemical-resistant
  • Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.
Specifications
  • Capacity:
    473 ml (16 oz.)
  • Dimensions:
    8.9 dia.×8.9H cm (3¹/₂×3¹/₂")