VWR® Microscope Slide Boxes for 25 Slides
Green
Catalog # 82003-420
Supplier: VWR International
Jump to:
Product Details & Documents
These heavy-duty, two-piece slide boxes feature cork lining and store up to 25 standard 25x75 mm (1x3") slides.
- Heavy-duty design
- Color-coded for easy identification
Boxes stack securely on top of each other. Ideal for storage or transport. Dimensions: 14Lx8.9Wx3.2Dcm (51/2x31/2x11/4").
Specifications
- Color:Green