Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Bacti Cell Spreaders

VWR® Bacti Cell Spreaders

Blue, 21.5 cm, 30 mm, Sterile
Catalog # 60828-682
Supplier:   VWR International
VWR® Bacti Cell Spreaders
Catalog # 60828-682
Supplier:  VWR International
VWR® Bacti Cell Spreaders
VWR® Bacti Cell Spreaders

Blue, 21.5 cm, 30 mm, Sterile

Price
$2,361.97
Log in for availability
Paquete de 25 Case de 4 (25UN)
Product Details & Documents

Convenient and economical cell spreaders are designed to simplify bacterial culture work.

  • Autoclavable
  • Simplify bacterial culture work
  • Durable fabrication

Durable enough to be autoclaved and reused.

 : Cell spreaders are available sterile (individually wrapped) or nonsterile (bulk packed).

Specifications
  • Sterility:
    Sterile
  • Color:
    Blue
  • Blade Width:
    30 mm
  • Overall Length:
    21.5 cm