VWR® Plug-Type Caps, PE
Green
Supplier: VWR International
VWR® Plug-Type Caps, PE
Catalog # 60819-080
Supplier: VWR International
Jump to:
Product Details & Documents
Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.
- Disposable
- Polyethylene construction
- Unique fluted design
- Snugly grasps plastic or glass tubes
They feature a unique fluted design that grips plastic or glass tubes snugly.
Ordering information: The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.
Specifications
- Color:Green
- Cat. No.:60819-080
- Environmentally Preferable:Sustainable Packaging|Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



