Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Spinbar® Micro Stir Bars

VWR® Spinbar® Micro Stir Bars

Stir Bar, Polygon, 12 mm (½"), 3 mm (1/8 "), PTFE
Catalog # 58948-397
Supplier:   VWR International
VWR® Spinbar® Micro Stir Bars
Catalog # 58948-397
Supplier:  VWR International
VWR® Spinbar®  Micro Stir Bars
VWR® Spinbar® Micro Stir Bars

Stir Bar, Polygon, 12 mm (½"), 3 mm (1/8 "), PTFE

Price
$29.90
Log in for availability
Each (1UN) Case de 50 (1UN)
Product Details & Documents

Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.

  • Particularly useful for environmental testing
  • Valuable for preparing and evaluating small sample volumes
  • Teflon® PTFE encapsulated
  • Autoclavable

Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.

Specifications
  • Coating:
    PTFE
  • Ø:
    3 mm (1/8 ")
  • Description:
    Stir Bar
  • Length:
    12 mm (½")
  • Shape:
    Polygon