VWR® Spinbar® Micro Stir Bars
Stir Bar, Polygon, 10 mm (⅜"), 3 mm (1/8 "), PTFE
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes, and gradient makers.
- Particularly useful for environmental testing
- Valuable for preparing and evaluating small sample volumes
- Teflon® PTFE encapsulated
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
- Coating:PTFE
- Ø:3 mm (1/8 ")
- Description:Stir Bar
- Length:10 mm (⅜")
- Shape:Polygon



