Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PIPET TIP N-STR ART100 PK1000
Catalog # 53225-738
Supplier:
MOLECULAR BIO-PRODUCTS INC
Expand image
PIPET TIP N-STR ART100 PK1000
Catalog # 53225-738
Supplier:
MOLECULAR BIO-PRODUCTS INC
Supplier Number:
2065B
Related Information
Documentation
Videos
Guide to ART Pipet Tips
Datasheet
Thermo Scientific Choosing the Best Pipette Tip
Thermo Scientific Control of Contamination
Documentation
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International