Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
55 mm, 45 mm, 10 mm, 59 mm
  30305-040
 :  4280-0550
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
  30305-040
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
Nalgene® Buchner Funnels, Polypropylene, Thermo Scientific
55 mm, 45 mm, 10 mm, 59 mm
$454.00
Each (1UN) Case de 8 (1UN)
Sturdy two-piece construction
Separable for easier cleaning. Highly resistant to most chemicals. Autoclavable.
  • Top I.D.:
    59 mm
  • Stem O.D.:
    10 mm
  • Stem Length:
    45 mm
  • Fits Paper Diameter:
    55 mm