Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

2000 ml, 10
Catalog # 29136-900
Supplier #:  4102-2000
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Catalog # 29136-900
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific
Nalgene® Erlenmeyer Flasks, Polypropylene, Thermo Scientific

2000 ml, 10

Price
$2,510.00
Log in for availability
Paquete de 2 Case de 3 (2UN)
Product Details & Documents

Wide-mouth flasks are built with heavy walls for hard use and a wide base for stability.

  • Liquid level easily visible
  • Autoclavable

The 2L flask has molded-in graduations.

Specifications
  • Capacity:
    2000 ml
  • Stopper No.:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.