PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
3 L, 10, 184 x 260 mm (71/4 x 101/4")
Supplier: Corning
: 4260-3L
Jump to:
Product Details & Documents
PYREX® borosilicate glass, clear, with short neck.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These flask mouth tooled for uniform rubber stopper fit. The normal working temperature is 230 °C.
Specifications
- Capacity:3 L
- Stopper No.:10
- O.D.×H:184 x 260 mm (71/4 x 101/4")



