Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flip Top Bottles

Flip Top Bottles

473 ml (16 oz.), Natural, 24-410, White PP unlined flip top, HDPE
Catalog # 16151-832
Supplier:   QORPAK
Supplier #:  PLC-03425
Flip Top Bottles
Catalog # 16151-832
Supplier:  QORPAK
Flip Top Bottles
Flip Top Bottles

473 ml (16 oz.), Natural, 24-410, White PP unlined flip top, HDPE

Price
$381.59
Log in for availability
Paquete de 6 Case de 4 (6UN)
Product Details & Documents

Flip top dispensing bottles are translucent and impact resistant.

  • Ideal for dispensing small amounts of liquid
  • Available in HDPE or LDPE formats

Dispensing bottles offer maximum squeezability.

Documents

Specifications
  • Cap Size:
    24-410
  • Cap Type:
    White PP unlined flip top
  • Capacity:
    473 ml (16 oz.)
  • Color:
    Natural
  • Material:
    HDPE