Order Entry
Mexico
Orders LinkContactUsLinkComponent
Mobile Phase Reservoir Bottles, Microsolv

Mobile Phase Reservoir Bottles, Microsolv

Mobile Phase Reservoir Bottle with Standard Screw Top Neck, 2 L (67.6 oz.)
Catalog # 97057-220
Supplier:   MICROSOLV TECH CORP
Supplier #:  58819-05R
Mobile Phase Reservoir Bottles, Microsolv
Catalog # 97057-220
Mobile Phase Reservoir Bottles, Microsolv
Mobile Phase Reservoir Bottles, Microsolv

Mobile Phase Reservoir Bottle with Standard Screw Top Neck, 2 L (67.6 oz.)

Price
$7,807.71
Log in for availability
Each (1UN)
Product Details & Documents

These borosilicate glass bottles are coated in an epoxy plastic.

Bottles will not break if dropped to the ground, even if full with solvent. They are available in the most commonly sized bottles, to be placed in the mobile phase tray of an HPLC system. These mobile phase reservoir bottles have either a 45 mm standard screw top neck, or a 29/32 Standard Taper Neck.

Ordering information: Each bottle is supplied with a drip ring and inert screw cap, if bottle has standard screw top neck.

Documents

Specifications
  • Capacity:
    2 L (67.6 oz.)
  • Description:
    Mobile Phase Reservoir Bottle with Standard Screw Top Neck