Order Entry
Mexico
Orders LinkContactUsLinkComponent
Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
7-Day Chart, –22...122 °F
Catalog # 97006-894
Supplier:   DICKSON COMPANY
Supplier #:  C206
Temperature Chart Recorders, Dickson®
Catalog # 97006-894
Supplier:  DICKSON COMPANY
Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
7-Day Chart, –22...122 °F
Price
$2,756.22
Log in for availability
Paquete de 60
Product Details & Documents
Recorders are ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability. Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers. Cord-free, portable, and easy to use, recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall. They feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door, and durable, corrosion-resistant black ABS and polycarbonate enclosure. Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times. They are °C/°F switchable.
  • Cord-free, portable, and easy to use
  • Recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall
  • Feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door
  • Durable, corrosion-resistant black ABS and polycarbonate enclosure
  • Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times
  • °C/°F switchable
Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers.
Average response time: 11 min. to move 63% of full scale
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. CE marked. IP21 rated.
Ordering information: Each recorder comes with a AA alkaline battery and a red pen. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Documents
Specifications
  • Temperature Range:
    –22...122 °F
  • Description:
    7-Day Chart
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in for availability
    Paquete de 6 - $2,985.88