Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Microscope Slide Holders/Mailers
VWR® Microscope Slide Holders/Mailers
82 x 39 mm (31/4 x 11/2"), Blue
Catalog # 95042-208
Supplier:   VWR International
VWR® Microscope Slide Holders/Mailers
Catalog # 95042-208
Supplier:  VWR International
VWR® Microscope Slide Holders/Mailers
VWR® Microscope Slide Holders/Mailers
82 x 39 mm (31/4 x 11/2"), Blue
Price
$956.37
Log in for availability
Case (250UN) Each (250UN) Cartón de 4 (250UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Single- or double-slide plastic holders/mailers feature sturdy crack-resistant construction and easy opening tabs.
  • Single- and double-slide formats available
  • Sturdy construction
  • Designed to accommodate standard slide sizes
Fit standard 25x75mm slides.
Specifications
  • Color:
    Blue
  • L×W:
    82 x 39 mm (31/4 x 11/2")