Order Entry
Mexico
Orders LinkContactUsLinkComponent
Wringers, Contec®

Wringers, Contec®

Flat Mop Wringer, PP, VertiKlean® MAX™ and QuickTask™ Mops (25 l (6.5 gal.) Buckets)
Catalog # 89198-854
Supplier:   CONTEC, INC. MX
Supplier #:  2738
Wringers, Contec®
Catalog # 89198-854
Supplier:  CONTEC, INC. MX
Wringers, Contec®
Wringers, Contec®

Flat Mop Wringer, PP, VertiKlean® MAX™ and QuickTask™ Mops (25 l (6.5 gal.) Buckets)

Price
$1,818.75
Log in for availability
Case (1UN)
Product Details & Documents

A variety of wringers designed for use with curved and flat mops.

  • Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
  • Can be used with curved, flat, and string mops
  • Fully autoclavable

These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.

Specifications
  • For Use With:
    VertiKlean® MAX™ and QuickTask™ Mops (25 l (6.5 gal.) Buckets)
  • Description:
    Flat Mop Wringer
  • Material:
    PP