$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
VWR BOTTLE WMPKR AMB 32OZ CS12
89043-120
:
DWK Life Sciences (KIMBLE)
VWR BOTTLE WMPKR AMB 32OZ CS12
89043-120
:
DWK Life Sciences (KIMBLE)
:
VW5223253V-21
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International