Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Dippers, PTFE

Dipper, 80 mm, 500 ml (16.7 oz.)
  89026-084
VWR® Dippers, PTFE
  89026-084
VWR® Dippers, PTFE
VWR® Dippers, PTFE

Dipper, 80 mm, 500 ml (16.7 oz.)

 
$7,206.80
   
Each (1UN)
 

Dippers feature a 60 cm (235/8") long steel shaft encapsulated in PTFE for rigidity.

  • Steel fabrication
  • Encapsulated in PTFE for rigidity
  • Sample most aggressive media confidently

Suitable for sampling the most aggressive media.

 : Dippers are supplied with one shaft; additional shafts (89026-222) are available to extend length.

 
  • Ø:
    80 mm
  • Capacity:
    500 ml (16.7 oz.)
  • Description:
    Dipper
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.