Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Dippers, PTFE

Dipper, 54 mm, 100 ml (3.33 oz.)
Catalog # 89026-080
Supplier:   VWR International
VWR® Dippers, PTFE
Catalog # 89026-080
Supplier:  VWR International
VWR® Dippers, PTFE
VWR® Dippers, PTFE

Dipper, 54 mm, 100 ml (3.33 oz.)

Price
$5,140.85
Log in for availability
Each (1UN)
Product Details & Documents

Dippers feature a 60 cm (235/8") long steel shaft encapsulated in PTFE for rigidity.

  • Steel fabrication
  • Encapsulated in PTFE for rigidity
  • Sample most aggressive media confidently

Suitable for sampling the most aggressive media.

 : Dippers are supplied with one shaft; additional shafts (89026-222) are available to extend length.

Specifications
  • Ø:
    54 mm
  • Capacity:
    100 ml (3.33 oz.)
  • Description:
    Dipper