Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® HDPE Multipurpose Containers
120 ml (4 oz.), 7 dia.×5.7H cm (2³/₄×2¹/₄")
  89009-662
VWR® HDPE Multipurpose Containers
  89009-662
VWR® HDPE Multipurpose Containers
VWR® HDPE Multipurpose Containers
120 ml (4 oz.), 7 dia.×5.7H cm (2³/₄×2¹/₄")
$2,002.29
Case (250UN)
Translucent containers for the storage of pathology, industrial, and environmental samples. Containers are made of high quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800mL container is equipped with a handle for portability.
  • HDPE construction
  • Chemical-resistant
  • Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.
  • Capacity:
    120 ml (4 oz.)
  • Dimensions:
    7 dia.×5.7H cm (2³/₄×2¹/₄")