$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
PIPET VOLUME BLU 1ML EA2 CS12
89003-474
:
DWK Life Sciences (KIMBLE)
PIPET VOLUME BLU 1ML EA2 CS12
89003-474
:
DWK Life Sciences (KIMBLE)
:
37000-1
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International