Flexi-Bin™ Carts, Labconco®
Product Details & Documents
For the safe transport of glassware and chemical containers
Carts have an 18-gauge tubular steel frame with a corrosion-resistant glacier white epoxy coating. Carts feature four 10.2cm (4") diameter casters (two with toe locks), two handles with ergonomic extruded rubber grips, and an identification tag. Carts support up to 181kg (400lbs.). Carts include one removable, corrosion-resistant polyethylene Flexi-Bin container and a bottom shelf to hold additional supplies.
Ordering information: Manufacturer's lifetime warranty. Shipped fully assembled. Also included are three long 51.4Lcm (201/4") and four short 38.7Lcm (151/4") polyethylene dividers for partitioning the bin to securely hold items in upright positions.
Documents
Labconco Carts Features
Videos
Specifications
- Description:Cart, 10 bins
- Dimensions:347/8x 19x363/8"
- Bin Dimensions:22.25×17"(e×t.);20.25×15"(int.)
- Shipping Weight:44 lbs.
- Load Capacity:181 kg (400 lbs.)
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



