Order Entry
Mexico
Orders LinkContactUsLinkComponent
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
7-Day Chart, 0...50 °C/°F or –50...0 °C/°F
Catalog # 61280-569
Supplier:   DICKSON COMPANY
Supplier #:  C653
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Catalog # 61280-569
Supplier:  DICKSON COMPANY
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
7-Day Chart, 0...50 °C/°F or –50...0 °C/°F
Price
$2,756.22
Log in for availability
Paquete de 60
Product Details & Documents
Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe. Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Increased shielding eliminates electronic noise interference and allows smoother pen movement.
Ordering information: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
  • Temperature Range:
    0...50 °C/°F or –50...0 °C/°F
  • Description:
    7-Day Chart
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in for availability
    Paquete de 6 - $2,985.88