Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Basic Protection SMS Lab Coats

Light blue, L, 40", 27", 32½"
  414004-355
VWR® Basic Protection SMS Lab Coats
  414004-355
VWR® Basic Protection SMS Lab Coats
VWR® Basic Protection SMS Lab Coats

Light blue, L, 40", 27", 32½"

 
$1,498.95
   
Case (30UN) Paleta de 25 (30UN)
 

VWR® Basic protection SMS lab coats are designed to provide lightweight, breathable coverage for non-hazardous environments. Constructed from SMS (Spunbond-Meltblown-Spunbond) material, they offer a reliable barrier against dust, dirt, and light liquid splashes while maintaining comfort during extended wear.

  • Lightweight, breathable, and durable SMS material for comfort during extended wear

  • Snap-front closure is easy to put on and remove while maintaining a secure fit
  • Collar design provides added coverage and a professional appearance
  • Shields against dust, dirt, and light liquid splashes in non-hazardous environments
  • Breathable fabric reduces heat buildup and improves wearer comfort over extended periods
  • Ideal for routine lab work and controlled environments without over-specifying protection

VWR® Basic protection SMS lab coats are engineered for environments where comfort and essential coverage are key. Constructed from SMS (Spunbond-Meltblown-Spunbond) fabric, these coats combine three layers of polypropylene to deliver a balance of strength, breathability, and barrier protection. The outer spunbond layers provide durability and resistance to tearing, while the meltblown middle layer acts as a filter against dust, dirt, and light liquid splashes. This makes them ideal for non-hazardous applications such as general laboratory work, cleanroom support tasks, and production areas where particulate control is important but chemical exposure is minimal.

Designed with practical features, these lab coats include a secure snap-front closure for easy donning and removal, a collar for added coverage, and elastic cuffs to ensure a snug fit that helps maintain cleanliness. Their lightweight construction promotes comfort during extended wear, reducing heat buildup and improving mobility. Available in multiple sizes, they support compliance with workplace safety protocols while maintaining a professional appearance. For facilities seeking cost-effective, disposable apparel that meets basic protection needs without sacrificing comfort, VWR® SMS lab coats offer a dependable solution.

 : Lab coats are packaged 30 per case.

 
  • Clothing Size:
    L
  • Color:
    Light blue
  • Length:
    40"
  • Chest Size:
    27"
  • Sleeve:
    32½"
  • Sleeve or leg length:
    Long
  • Gender:
    Unisex
  • Closure type:
    Press studs / snaps
  • Water resistance:
    16.7 G
  • Tensile strength:
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • Weight:
    50.45 g/m²
  • Bacterial filtration efficiency:
    90%
  • Tearing strength:
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • Cat. No.:
    414004-355
  • Cleanroom class:
    10000/ISO 7
  • Air permeability:
    95.3 cu. ft./min./sq. ft.
  • Garment application:
    Cleanroom|Industrial|Laboratory
  • Pockets:
    With pockets
  • Fabric:
    Spunbound polypropylene
  • Particle shedding (Helmke Drum):
    Level II
  • Water vapor transmission rate:
    965 g/m²/24 h