Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

6 L, 11
Catalog # 29111-208
Supplier:   Corning
Número de proveedor #:  4060-6L
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 29111-208
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

6 L, 11

Price
$5,040.00
Log in for availability
Case (1UN)
Product Details & Documents

Manufactured from borosilicate glass.

  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Specifications
  • Capacity:
    6 L
  • Stopper No.:
    11