Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
2 L, 10
Catalog # 28196-373
Supplier: Thermo Fisher Scientific
: DS4101-2000
Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
Catalog # 28196-373
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
One-piece molded for strength and stability
Withstand full vacuum to 762mm Hg (30in. Hg). Tubulation at 45° angle prevents tipping. Oval marking area. Barbed adapter accepts 6.4mm (1/4") to 9.5mm (3/8") I.D. tubing. Autoclavable.
Specifications
- Capacity:2 L
- Rubber Stopper Size:10