Order Entry
Mexico
Orders LinkContactUsLinkComponent
LabSolutions™ Detergents, Labconco®
LabSolutions™ Non-Foaming Powder Detergent, bucket, 4.5 kg (10 lbs.)
  21851-130
 :  4422000
LabSolutions™ Detergents, Labconco®
  21851-130
LabSolutions™ Detergents, Labconco®
LabSolutions™ Detergents, Labconco®
LabSolutions™ Non-Foaming Powder Detergent, bucket, 4.5 kg (10 lbs.)
$3,730.00
Each (10lb)
Formulated for use in automatic laboratory glassware washers
Low-Foaming Liquid Detergent removes a wide range of laboratory contaminants. Noncorrosive formula is safe for use on stainless steel, glass, or plastic. Phosphate-free for environmentally safe disposal and chlorine-free for safe use tissue cultures. Can be used in hot or cold water. Comes in a 3.8L (1gal.) jug. Washes approximately 180 loads.
Non-Foaming Powder Detergent effectively removes grease, blood, agar, and protein digestates from glassware. Special formula prevents damage to machine pumps, seals, and gaskets. A 1% solution has an approximate pH of 12.5. Available in a 4.5kg (10lbs.) or 12.5kg (27.5lbs.) bucket.
Neutralizing Acid Rinse removes hard water deposits, such as mineral deposits, accumulated salts, and lime scale. Mild acid corrects pH, reducing alkaline detergent residue. Comes in a 1L (32oz.) bottle. Fills reservoir approximately six times.
Specially designed for the Labconco® SteamScrubber® and FlaskScrubber® washers. Available in liquid, powder, and a neutralizing acid rinse. All formulations rinse clean away.
  • Description:
    LabSolutions™ Non-Foaming Powder Detergent, bucket
  • Size:
    4.5 kg (10 lbs.)