VWR® Burets, Automatic Self-Zeroing, Class A, with Stopcock
10 ml, 0.05 ml, 1000 ml, 24/40, Fluoropolymer resin
17580-010
Borosilicate glass buret assembly with precision-grade white scale.
- Specially designed for accuracy and economy
Assembly seats securely into [ST] ground glass bottle top. Manufactured to Class A specifications for burets.
: Shipped complete with bottle, rubber bulb, and stopcock.
- Graduation Interval:0.05 ml
- ST Joint No.:24/40
- Capacity:10 ml
- Reservoir Capacity:1000 ml
- Stopcock Type:Fluoropolymer resin



