Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polypropylene Snap-Seal™ Sample Containers, Corning®
Polypropylene Snap-Seal™ Sample Containers, Corning®
110 ml (3.7 oz.), Low-Profile Snap-Seal™ Sample Containers
Catalog # 16126-022
Supplier:   Corning
Supplier #:  1730-4L
Polypropylene Snap-Seal™ Sample Containers, Corning®
Catalog # 16126-022
Supplier:  Corning
Polypropylene Snap-Seal™ Sample Containers, Corning®
Polypropylene Snap-Seal™ Sample Containers, Corning®
110 ml (3.7 oz.), Low-Profile Snap-Seal™ Sample Containers
Price
$4,060.00
Log in for availability
Case (200UN)
Product Details & Documents
Designed for a wide variety of applications, these containers provide a reliable leak-tight seal when closed properly.
  • Snap-Seal locking device keeps the cap closed and secure
  • Hinged cap reduces the chance of sample contamination
  • Graduations in both millilitres and ounces
  • Rough source for marking
The 120mL (4oz.) size is available in both high- and low-profile design.
Specifications
  • Capacity:
    110 ml (3.7 oz.)
  • Description:
    Low-Profile Snap-Seal™ Sample Containers